CAS RN 62956-48-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Gomisin G, MedChemExpress
MedChemExpress Gomisin G is an ethanolic extract of the stems of Kadsura interior; exhibits potent anti-HIV activity with EC50 and therapeutic index (TI) values of 0.006 microgram/mL and 300, respectively.