CAS RN 625110-37-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DPP-IV-IN-1, MedChemExpress
MedChemExpress DPP-IV-IN-1 is a potent inhibitor of dipeptidyl peptidase IV (DPP-IV), a highly specific serine protease, with an IC50 of 4.6 nM.