CAS RN 61596-47-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Prolylleucine
MedChemExpress Prolylleucine is a dipeptide containing branched-chain amino acids. Prolylleucine can affect the circadian rhythms and behaviour of animals.
2
MedChemExpress Prolylleucine
MedChemExpress Prolylleucine is a dipeptide containing branched-chain amino acids. Prolylleucine can affect the circadian rhythms and behaviour of animals.
3
MedChemExpress Prolylleucine
MedChemExpress Prolylleucine is a dipeptide containing branched-chain amino acids. Prolylleucine can affect the circadian rhythms and behaviour of animals.