CAS RN 6151-40-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Quinidine hydrochloride monohydrate, MedChemExpress
MedChemExpress Quinidine hydrochloride monohydrate is an anti-arrythmic agent which is also a potent blocker of K+ channel with an IC50 of 19.9 μM.