CAS RN 613225-56-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
F1063-0967, MedChemExpress
MedChemExpress F1063-0967 is a Dual-specificity phosphatase 26 (DUSP26) inhibitor with an IC50 of 11.62 μM.