CAS RN 612530-44-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PTP1B-IN-1, MedChemExpress
MedChemExpress PTP1B-IN-1 is a potent protein tyrosine phosphatase-1B (PTP1B) inhibitor with IC50 of 1.6 mM; 1,2,5-thiadiazolidin-3-one-1,1-dioxide scaffold for derivatives synthesis.