CAS RN 6106-07-6
IUPAC Nombre:
sodium (2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoate pentahydrate
Sinónimos:
sodium levothyroxine(1-) pentahydrate
Peso molecular (g/mol):
888.93
Fórmula molecular:
C15H20I4NNaO9
InChi Key:
JMHCCAYJTTWMCX-QWPJCUCISA-M
SMILES:
O.O.O.O.O.[Na+].N[C@@H](CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C([O-])=O
1
–
1
de
1
resultados
1
L-Thyroxine sodium salt pentahydrate, MedChemExpress
MedChemExpress L-Thyroxine sodium salt pentahydrate (Levothyroxine; T4) is a synthetic hormone for the research of hypothyroidism. DIO enzymes convert biologically active thyroid hormone (Triiodothyronine,T3) from L-Thyroxine (T4).