CAS RN 607373-46-6
IUPAC Nombre:
4-[7-(3-aminopyrrolidine-1-carbonyl)-1-ethyl-1H-imidazo[4,5-c]pyridin-2-yl]-1,2,5-oxadiazol-3-amine dihydrochloride
Sinónimos:
4-[7-(3-aminopyrrolidine-1-carbonyl)-1-ethylimidazo[4,5-c]pyridin-2-yl]-1,2,5-oxadiazol-3-amine dihydrochloride
Peso molecular (g/mol):
415.28
Fórmula molecular:
C15H20Cl2N8O2
InChi Key:
HSROPTGRSYJPJY-UHFFFAOYNA-N
SMILES:
Cl.Cl.CCN1C(=NC2=CN=CC(C(=O)N3CCC(N)C3)=C12)C1=NON=C1N
1
–
2
de
2
resultados
1
Tocris Bioscience™ SB 772077B dihydrochloride
CAS: 607373-46-6 Fórmula molecular: C15H20Cl2N8O2
2
SB-772077B dihydrochloride, MedChemExpress
MedChemExpress SB-772077B dihydrochloride is an aminofurazan-based Rho kinase (ROCK) inhibitor with IC50s of 5.6 nM and 6 nM toward ROCK1 and ROCK2, respevtively.