CAS RN 60285-94-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
H-Arg-4MβNA, MedChemExpress
MedChemExpress H-Arg-4MβNA is a substrate for cathepsin H, used for the detection of enzyme activity in gel electrophoresis.