CAS RN 587850-67-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
C-7280948, MedChemExpress
MedChemExpress C-7280948 is a selective and potent protein methyltransferase1 (PRMT1) inhibitor with an IC50 value of 12.75 μM.