CAS RN 5853-29-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Cephaeline dihydrochloride, MedChemExpress
MedChemExpress Cephaeline dihydrochloride is a selective CYP2D6 inhibtor with an IC50 of 121 μM.