CAS RN 581073-80-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
Tocris Bioscience™ KJ Pyr 9
High affinity Myc inhibitor
2
KJ Pyr 9, MedChemExpress
MedChemExpress KJ Pyr 9 is an inhibitor of MYC with a Kd of 6.5 nM in in vitro assay.
3
4