CAS RN 58-49-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
MedChemExpress Angiotensin II 5-valine
MedChemExpress Angiotensin II 5-valine is an agonist of angiotensin receptor.
2
MedChemExpress Angiotensin II 5-valine
MedChemExpress Angiotensin II 5-valine is an agonist of angiotensin receptor.
3
MedChemExpress Angiotensin II 5-valine
MedChemExpress Angiotensin II 5-valine is an agonist of angiotensin receptor.
4
MedChemExpress Angiotensin II 5-valine
MedChemExpress Angiotensin II 5-valine is an agonist of angiotensin receptor.