CAS RN 572924-54-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Ridaforolimus, >80%, TRC
CAS: 572924-54-0 Fórmula molecular: C53 H84 N O14 P Peso molecular (g/mol): 990.21 Sinónimo: Rapamycin, 42-(dimethylphosphinate) (9CI),AP 23573,Deforolimus,MK 8669,Ridaforolimus SMILES: CO[C@@H]1C[C@H](C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(/C)\[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C(/C)\[C@H](C[C@@H]3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N4CCCC[C@H]4C(=O)O2)OC)CC[C@H]1OP(=O)(C)C
2
Ridaforolimus, MedChemExpress
MedChemExpress Ridaforolimus (MK-8669) is a potent and selective mTOR inhibitor; inhibits ribosomal protein S6 phosphorylation with an IC50 of 0.2 nM in HT-1080 cells.