CAS RN 57046-73-8
IUPAC Nombre:
2-(1H-indol-3-yl)-3-(pyridin-3-yl)prop-2-enenitrile
Sinónimos:
2-(1H-indol-3-yl)-3-(pyridin-3-yl)prop-2-enenitrile
Peso molecular (g/mol):
245.29
Fórmula molecular:
C16H11N3
InChi Key:
YMYYQRZCCKBFBE-UHFFFAOYSA-N
SMILES:
N#CC(=CC1=CN=CC=C1)C1=CNC2=CC=CC=C12
1
–
1
de
1
resultados
1
Paprotrain, MedChemExpress
MedChemExpress Paprotrain is a cell-permeable inhibitor of the kinesin MKLP-2, inhibits the ATPase activity of MKLP-2 with an IC50 of 1.35 μM and a Ki of 3.36 μM and shows a moderate inhibition activity on DYRK1A with an IC50 of 5.5 μM.