CAS RN 5699-54-7
IUPAC Nombre:
(3R)-3-azaniumyl-4-methylpentanoate
Sinónimos:
β-leucine zwitterion (3R)-β-leucine zwitterion
Peso molecular (g/mol):
131.18
Fórmula molecular:
C6H13NO2
InChi Key:
GLUJNGJDHCTUJY-RXMQYKEDSA-N
SMILES:
CC(C)[C@H]([NH3+])CC([O-])=O
1
–
1
de
1
resultados
1
3-Amino-4-methylpentanoic acid, MedChemExpress
MedChemExpress 3-Amino-4-methylpentanoic acid is a beta amino acid and positional isomer of L-leucine which is naturally produced in humans via the metabolism of L-leucine by the enzyme leucine 2,3-aminomutase.