CAS RN 56605-16-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NCI172112, MedChemExpress
MedChemExpress NCI172112 is a classical bifunctional alkylating agent synthesized in an effort to develop antitumor agents effective against CNS tumors.