CAS RN 565460-15-3
IUPAC Nombre:
cyclohexyl N-{[1,1'-biphenyl]-3-yl}carbamate
Sinónimos:
cyclohexyl N-{[1,1'-biphenyl]-3-yl}carbamate
Peso molecular (g/mol):
295.38
Fórmula molecular:
C19H21NO2
InChi Key:
HHVUFQYJOSFTEH-UHFFFAOYSA-N
SMILES:
O=C(NC1=CC=CC(=C1)C1=CC=CC=C1)OC1CCCCC1
1
–
1
de
1
resultados
1
URB602, MedChemExpress
MedChemExpress URB602 is a selective monoacylglycerol lipase (MGL) inhibitor, which inhibits rat brain MGL with IC50 of 28±4 μM through a noncompetitive mechanism.