CAS RN 5614-64-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
8-Hydroxyguanine, MedChemExpress
MedChemExpress 8-Hydroxyguanine is a major pre-mutagenic lesion generated from reactive oxygen species. It causes G-T and A-C substitutions.