CAS RN 56100-42-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
l-Atabrine dihydrochloride, MedChemExpress
MedChemExpress l-Atabrine dihydrochloride is a less active enantiomer of quinacrine which displays antiprion activity.