CAS RN 56100-41-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
d-Atabrine dihydrochloride, MedChemExpress
MedChemExpress d-Atabrine dihydrochloride is an active enantiomer of quinacrine which displays antiprion activity.