CAS RN 5610-40-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Securinine, TRC
CAS: 5610-40-2 Fórmula molecular: C13 H15 N O2 Peso molecular (g/mol): 217.26 SMILES: O=C1O[C@@]23C[C@@H](C=CC2=C1)N4CCCC[C@H]34
2
(-)-Securinine, MedChemExpress
MedChemExpress (-)-Securinine is plant-derived alkaloid and also a GABAA receptor antagonist.