CAS RN 56049-88-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Indacrinone, MedChemExpress
MedChemExpress Indacrinone (MK196) is an investigational diuretic which has pronounced saluretic activity in the rat and dog as well as both uricosuric and saluretic activity in the chimpanzee.