CAS RN 5588-20-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Chlordantoin, MedChemExpress
MedChemExpress Chlordantoin is an antifungal agent and has the potential for vaginal candidiasis treatment.