CAS RN 55482-89-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Guacetisal, MedChemExpress
MedChemExpress Guacetisal is obtained from the esterification of acetylsalicylic acid with guaiacol which has the potential for chronic bronchitis treatment extracted from patent CN 106866420 A.