CAS RN 55476-47-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PSB-12062, MedChemExpress
MedChemExpress PSB-12062 is a potent and selective P2X4 antagonist with an IC50 of 1.38 μM for human P2X4.