CAS RN 55285-14-8
IUPAC Nombre:
2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl N-[(dibutylamino)sulfanyl]-N-methylcarbamate
Sinónimos:
carbosulfan caswell no. 463C marshal dibutylaminosulfenylcarbofuran marshall posse epa pesticide chemical code 090602
Peso molecular (g/mol):
380.55
Fórmula molecular:
C20H32N2O3S
InChi Key:
JLQUFIHWVLZVTJ-UHFFFAOYSA-N
SMILES:
CCCCN(CCCC)SN(C)C(=O)OC1=CC=CC2=C1OC(C)(C)C2
1
–
3
de
3
resultados
1
Carbosulfán, TRC
CAS: 55285-14-8 Fórmula molecular: C20 H32 N2 O3 S Peso molecular (g/mol): 380.54 Sinónimo: Carbamic acid, [(dibutylamino)thio]methyl-, 2,3-dihydro-2,2-dimethyl-7-benzofuranyl ester (9CI),Advantage,Carbosulfan,DBSC,Dingliukebaiwei,FMC 35001,Marshal,Marshal 25EC,Marshal 25ST,Posse,Versal,Zaprawa Marshal Nombre IUPAC: (2,2-dimetil-3H-1-benzofurano-7-il) N-(dibutilamino)sulfanil-N-metilcarbamato SMILES: CCCCN(CCCC)SN(C)C(=O)Oc1cccc2CC(C)(C)Oc12
2
Carbosulfan, MedChemExpress
MedChemExpress Carbosulfan inhibited relatively potently CYP3A4 and moderately CYP1A1/2 and CYP2C19 in pooled HLM (human livers).