CAS RN 552325-16-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
A-443654, MedChemExpress
MedChemExpress A-443654 is a pan-Akt inhibitor and has equal potency against Akt1, Akt2, or Akt3 within cells (Ki=160 pM).