CAS RN 546111-97-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Litronesib Racemate, MedChemExpress
MedChemExpress Litronesib Racemate (LY2523355 Racemate) is the racemate of litronesib. Litronesib is a selective, allosteric inhibitor of kinesin Eg5.