CAS RN 544467-07-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HIV-1 integrase inhibitor, MedChemExpress
MedChemExpress HIV-1 integrase inhibitor is uesful for anti-HIV.