CAS RN 53947-92-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Corylin, MedChemExpress
MedChemExpress Corylin is a major bioactive compound isolated from Psoralea corylifolia L; antibiotic or anticancer compound.