CAS RN 53868-26-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(Rac)-NSC305787 hydrochloride, MedChemExpress
MedChemExpress NSC305787 hydrochloride is an inhibitor of ezrin with a Kd of 5.85 μM, inhibits the phosphorylation of ezrin caused by PKCι with an IC50 of 8.3 μM, has antitumor activity.