CAS RN 538-23-8
IUPAC Nombre:
1,3-bis(octanoyloxy)propan-2-yl octanoate
Sinónimos:
Rato maceight trioctonoin trioctanoylglycerol octanoin, tri- (8CI) glyceryl tricaprylate glyceryl trioctanoate sefsol 800 tricaprylin tricaprylic glyceride
Peso molecular (g/mol):
470.69
Fórmula molecular:
C27H50O6
InChi Key:
VLPFTAMPNXLGLX-UHFFFAOYSA-N
SMILES:
CCCCCCCC(=O)OCC(COC(=O)CCCCCCC)OC(=O)CCCCCCC
1
–
1
de
1
resultados
1
Tricaprilin, MedChemExpress
MedChemExpress Tricaprilin (Trioctanoin) is used in study for patients with mild to moderate Alzheimer's disease and has a role as an anticonvulsant and a plant metabolite.