CAS RN 52949-83-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Ajugol, MedChemExpress
MedChemExpress Ajugol is an iridoid glycoside that can be isolated from Sideritis germanicopolitana. Ajugol has anti-protozoal activity againt Trypanosoma b. rhodesiense with an IC50 of 31.8 μg/mL.