CAS RN 524923-88-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
JNJ-10229570, MedChemExpress
MedChemExpress JNJ-10229570 is an antagonist of melanocortin receptor 1 (MC1R) and melanocortin receptor 5 (MC5R), which inhibits sebaceous gland differentiation and the production of sebum-specific lipids. JNJ-10229570 inhibits the binding of 125I-NDP-α-MSH to cells expressing human MC1R and MC5R, with IC50 values of 270 nM and 200 nM, respectively.