CAS RN 52452-60-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Terphenyllin, MedChemExpress
MedChemExpress Terphenyllin is a naturally abundant p-terphenyl metabolite isolated from the coral derived fungus Aspergillus candidus, has significant α-glucosidase inhibitory activity.