CAS RN 524-30-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Fraxin, MedChemExpress
MedChemExpress Fraxin isolated from Acer tegmentosum, F. ornus or A. hippocastanum, is a glucoside of fraxetin and reported to exert potent anti-oxidative stress action, anti-inflammatory and antimetastatic properties. Fraxin shows its antioxidative effect through inhibition of cyclo AMP phosphodiesterase enzyme.