CAS RN 51644-96-3
IUPAC Nombre:
tert-butyl N-(5-aminopentyl)carbamate
Sinónimos:
tert-butyl N-(5-aminopentyl)carbamate
Peso molecular (g/mol):
202.3
Fórmula molecular:
C10H22N2O2
InChi Key:
DPLOGSUBQDREOU-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NCCCCCN
1
–
1
de
1
resultados
1
NH2-C5-NH-Boc, MedChemExpress
MedChemExpress NH2-C5-NH-Boc (PROTAC Linker 23) is an alkyl chain-based PROTAC linker can be used in the synthesis of PROTACs.