CAS RN 5128-44-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
7,4'-Di-O-methylapigenin, MedChemExpress
MedChemExpress The compound 7,4'-Di-O-methylapigenin may be partly responsible for the reported antifungal activity of C.