CAS RN 5081-48-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
MedChemExpress Ellipticine hydrochloride
MedChemExpress Ellipticine (NSC 71795) hydrochloride is a potent antineoplastic agent; inhibits DNA topoisomerase II activities.
2
MedChemExpress Ellipticine hydrochloride
MedChemExpress Ellipticine (NSC 71795) hydrochloride is a potent antineoplastic agent; inhibits DNA topoisomerase II activities.
3
MedChemExpress Ellipticine hydrochloride
MedChemExpress Ellipticine (NSC 71795) hydrochloride is a potent antineoplastic agent; inhibits DNA topoisomerase II activities.
4
MedChemExpress Ellipticine hydrochloride
MedChemExpress Ellipticine (NSC 71795) hydrochloride is a potent antineoplastic agent; inhibits DNA topoisomerase II activities.