CAS RN 50667-69-1
IUPAC Nombre:
2,2,2-trifluoro-N-(hydroxymethyl)acetamide
Sinónimos:
2,2,2-trifluoro-N-(hydroxymethyl)acetamide
Peso molecular (g/mol):
143.07
Fórmula molecular:
C3H4F3NO2
InChi Key:
URUWXKFAEKTWKG-UHFFFAOYSA-N
SMILES:
OCNC(=O)C(F)(F)F
1
–
1
de
1
resultados
1
N-(Hidroximetil)trifluoroacetamida, + 98 %, Thermo Scientific Chemicals
CAS: 50667-69-1 Fórmula molecular: C3H4F3NO2 Peso molecular (g/mol): 143.065 Número MDL: MFCD00014416 Clave InChI: URUWXKFAEKTWKG-UHFFFAOYSA-N Sinónimo: 2,2,2-trifluoro-n-hydroxymethyl acetamide,n-hydroxymethyl trifluoroacetamide,n-hydroxymethyltrifluoroacetamide,acetamide, 2,2,2-trifluoro-n-hydroxymethyl,acmc-20akaf,trifluoroacetamidomethanol,n-hydroxymethyl fluoroacetamide,n-hydroxymethyl-trifluoroacetamide,n-hydroxy methyl trifluoro acetamide PubChem CID: 3084931 Nombre IUPAC: 2,2,2-trifluoro-N-(hidroximetil)acetamida SMILES: C(NC(=O)C(F)(F)F)O