CAS RN 50264-69-2
IUPAC Nombre:
1-[(2,4-dichlorophenyl)methyl]-1H-indazole-3-carboxylic acid
Sinónimos:
DICA LND 氯尼达明 lonidaminum lonidamine lonidamina lonidamin doridamina diclondazolic acid
Peso molecular (g/mol):
321.16
Fórmula molecular:
C15H10Cl2N2O2
InChi Key:
WDRYRZXSPDWGEB-UHFFFAOYSA-N
SMILES:
OC(=O)C1=NN(CC2=CC=C(Cl)C=C2Cl)C2=CC=CC=C12
1
–
2
de
2
resultados
1
Tocris Bioscience™ Lonidamine
CAS: 50264-69-2 Fórmula molecular: C15H10Cl2N2O2
2
Lonidamine, MedChemExpress
MedChemExpress Lonidamine (AF-1890) is a hexokinase and mitochondrial pyruvate carrier inhibitor (Ki: 2.5 μM). Lonidamine also inhibits aerobic glycolysis in cancer cells. Lonidamine can be used in the research of mitochondrial metabolism and inflammation, such as pulmonary fibrosis.