CAS RN 501925-31-1
IUPAC Nombre:
1-(5-chloro-2,4-dimethoxyphenyl)-3-(5-methyl-1,2-oxazol-3-yl)urea
Sinónimos:
1-(5-chloro-2,4-dimethoxyphenyl)-3-(5-methyl-1,2-oxazol-3-yl)urea
Peso molecular (g/mol):
311.72
Fórmula molecular:
C13H14ClN3O4
InChi Key:
CEIIEALEIHQDBX-UHFFFAOYSA-N
SMILES:
COC1=CC(OC)=C(Cl)C=C1NC(=O)NC1=NOC(C)=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ PNU 120596
Positive allosteric modulator of α7 nAChRs; active in vivo
2
PNU-120596, MedChemExpress
MedChemExpress PNU-120596 (NSC 216666) is a potent and selective α7 nAChR positive allosteric modulator (PMA) with an EC50 of 216 nM. PNU-120596 is inactive against α4β2, α3β4, and α9α10 nAChRs. PNU-120596 has the potential for psychiatric and neurological disorders research.