CAS RN 488847-28-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
ORM-10103, MedChemExpress
MedChemExpress ORM-10103 is a specific inhibitor of the Na+/Ca2+ exchanger (NCX), which decreases the NCX current with estimated IC50s of 55 and 67 nM at -80 and at 20 mV, respectively.