CAS RN 4846-19-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
N-Nornuciferine, MedChemExpress
MedChemExpress N-Nornuciferine is an aporphine alkaloid in lotus leaf that significantly inhibits CYP2D6 with IC50 and Ki of 3.76 and 2.34 μM, respectively.