CAS RN 478963-79-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Calcium-Sensing Receptor Antagonists I, MedChemExpress
MedChemExpress Calcium-Sensing Receptor Antagonists I is an antagonist of calcium-sensing parathyroid hormone receptors.