CAS RN 476310-60-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
VO-Ohpic trihydrate, MedChemExpress
MedChemExpress VO-Ohpic trihydrate is a highly potent inhibitor of PTEN with an IC50 of 46±10 nM.