CAS RN 476159-98-5
IUPAC Nombre:
N-(2-chloro-5-methoxyphenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
Sinónimos:
N-(2-chloro-5-methoxyphenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
Peso molecular (g/mol):
442.94
Fórmula molecular:
C23H27ClN4O3
InChi Key:
WPOXAFXHRJYEIC-UHFFFAOYSA-N
SMILES:
COC1=CC(NC2=NC=NC3=CC(OCC4CCN(C)CC4)=C(OC)C=C23)=C(Cl)C=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ AZM 475271
CAS: 476159-98-5 Fórmula molecular: C23H27ClN4O3
2
AZM475271, MedChemExpress
MedChemExpress AZM475271 is a potent and selective Src kinase inhibitor with IC50 of 5 nM; no inhibitory activity on Flt3, KDR, Tie-2.