CAS RN 4551-16-0
IUPAC Nombre:
trimethyl[(trimethylsilyl)tellanyl]silane
Sinónimos:
trimethyl[(trimethylsilyl)tellanyl]silane
Peso molecular (g/mol):
273.98
Fórmula molecular:
C6H18Si2Te
InChi Key:
VMDCDZDSJKQVBK-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)[Te][Si](C)(C)C