CAS RN 439685-79-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Pro-xylane, MedChemExpress
MedChemExpress Pro-xylane (Hydroxypropyl tetrahydropyrantriol) is a biologically active C-glycoside in aqueous media, acts as an activator of glycosaminoglycans (GAGs) biosynthesis. Pro-xylane is the first example of 'Green' chemical used in cosmetic.